Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethyl-3-hydroxy-6-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. E08335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E08335G Supplier TCI America Supplier No. E08335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity (min. 98.0%) pyridine derivative used for laboratory research purposes.

Chemical Identifiers

CAS 2364-75-2
Molecular Formula C8H12ClNO
Molecular Weight (g/mol) 173.64
MDL Number MFCD00462409
InChI Key KZUIXWYHQJZUOK-UHFFFAOYSA-N
Synonym 2-Ethyl-6-methyl-3-pyridinol, Emoxypine
PubChem CID 114681
IUPAC Name hydrogen 2-ethyl-6-methylpyridin-3-ol chloride
SMILES [H+].[Cl-].CCC1=NC(C)=CC=C1O

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 5 g
Formula Weight 137.18
Percent Purity ≥98.0% (GC,T)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material 2-Ethyl-3-hydroxy-6-methylpyridine
RTECSNumber UU7714800
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.