Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-((ethyl(4-fluorophenyl)amino)methyl)-7,8-dihydro-4H,6H-cyclopentathiazolo[3,2-a]pyrimidin-4-one | 1698901-76-6 | 98.9% | 343.42 g·mol⁻¹ | C18H18FN3OS | 1 ML
SDP

Catalog No. 5000240609
Encompass_Preferred

GNE-8324 is a selective GluN2A positive allosteric modulator of NMDA receptors. It selectively enhances NMDAR-mediated synaptic responses in inhibitory neurons but not excitatory neurons, and is used as a research tool to probe GluN2A-dependent receptor function.

  • Selective GluN2A positive allosteric modulator
  • Enhances NMDAR-mediated synaptic responses in inhibitory neurons
  • Useful tool for studying GluN2A-dependent NMDA receptor pharmacology
  • Offered as a 10 mM solution in DMSO (1 mL) and as solid quantities
  • High purity, approximately 98.9%
  • Molecular formula C18H18FN3OS and molecular weight 343.42 g·mol⁻¹
  • Storage recommendations provided for powder and solvent to preserve stability

Catalog No. 50-002-40609 Supplier Medchemexpress LLC Supplier No. HY10749810MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GNE-8324 is a selective GluN2A positive allosteric modulator of NMDA receptors. It selectively enhances NMDAR-mediated synaptic responses in inhibitory neurons but not excitatory neurons, and is used as a research tool to probe GluN2A-dependent receptor function.

  • Selective GluN2A positive allosteric modulator
  • Enhances NMDAR-mediated synaptic responses in inhibitory neurons
  • Useful tool for studying GluN2A-dependent NMDA receptor pharmacology
  • Offered as a 10 mM solution in DMSO (1 mL) and as solid quantities
  • High purity, approximately 98.9%
  • Molecular formula C18H18FN3OS and molecular weight 343.42 g·mol⁻¹
  • Storage recommendations provided for powder and solvent to preserve stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.