Learn More
Medchemexpress LLC 2-Ethyl-6-methylaniline-d13 | 1219794-93-0 | 98.6% | 148.29 | 10 MG
SDP
Catalog No. 5000453879
Encompass_Preferred
2-Ethyl-6-methylaniline-d13 is the deuterium labeled 2-Ethyl-6-methylaniline. It is designed for research use only and not for patient use. This compound serves as a tracer and an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS. Incorporating stable heavy isotopes into drug molecules, particularly deuterium, offers the potential to influence pharmacokinetic and metabolic profiles of drugs during development.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling can impact pharmacokinetic and metabolic profiles of pharmaceuticals
Catalog No. 50-004-53879 Supplier Medchemexpress LLC Supplier No. HYW012626S10MG
2-Ethyl-6-methylaniline-d13 is the deuterium labeled 2-Ethyl-6-methylaniline. It is designed for research use only and not for patient use. This compound serves as a tracer and an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS. Incorporating stable heavy isotopes into drug molecules, particularly deuterium, offers the potential to influence pharmacokinetic and metabolic profiles of drugs during development.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling can impact pharmacokinetic and metabolic profiles of pharmaceuticals