Learn More
Medchemexpress LLC 2-Ethyl-6-methylaniline-d13 | 1219794-93-0 | 98.6% | 148.29 | 100 MG
SDP
Catalog No. 5000366580
Encompass_Preferred
2-Ethyl-6-methylaniline-d13 is the deuterium labeled 2-Ethyl-6-methylaniline. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-66580 Supplier Medchemexpress LLC Supplier No. HYW012626S100MG
2-Ethyl-6-methylaniline-d13 is the deuterium labeled 2-Ethyl-6-methylaniline. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs