Learn More
Medchemexpress LLC HY-W089538 500mg Medchemexpress, 2-Ethyl-6-methylphenol CAS:1687-64-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW089538500MG
Encompass_Preferred
Medchemexpress, HY-W089538 500mg 2-Ethyl-6-methylphenol CAS:1687-64-5 2-Ethyl-6-methylphenol, an alkylphenol, is isolated form the tumorigenic neutral subfraction of cigarette smoke condensate. 2-Ethyl-6-methylphenol exhibits insecticidal and bactericidal activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3510