Learn More
Cayman Chemical 2-Ethylamino-1-phenylbutane (hydrochloride), Purity: ≥98%, CAS: 29805-52-5, 50 mg
SDP
Catalog No. 502894701
Encompass_Preferred
An analog of N-ethylamphetamine that is characterized by an ethyl group replacing methyl at the α position
Catalog No. 50-289-4701 Supplier Cayman Chemical Supplier No. 900186350 MG
An analog of N-ethylamphetamine that is characterized by an ethyl group replacing methyl at the α position