Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylbenzeneboronic acid, 98+%, Thermo Scientific™

Catalog No. AAL1771903
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1771903 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1771903 Supplier Thermo Scientific Chemicals Supplier No. L1771903

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 90002-36-1
Molecular Formula C8H11BO2
Quantity 1g
MDL Number MFCD02093075
Beilstein 2963801
Synonym 2-ethylbenzeneboronic acid, 2-ethylphenyl boronic acid, 2-ethylphenyl boranediol, boronic acid, 2-ethylphenyl, 2-ethylphenylboronicacid, o-ethylphenylboronic acid, pubchem14050, 2-ethylphenylboranic acid, 2-ethylbenzene boronic acid
InChI Key QSSPYZOSTJDTTL-UHFFFAOYSA-N
SMILES CCC1=CC=CC=C1B(O)O
IUPAC Name (2-ethylphenyl)boronic acid
Molecular Weight (g/mol) 149.98
PubChem CID 4100862
Formula Weight 149.99
Percent Purity ≥98%
Chemical Name or Material 2-Ethylbenzeneboronic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 677-274-9
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.