Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Ethylbutyraldehyde

Catalog No. 110094250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-009-4250ML 250 mL
11-009-41L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-009-4250ML Supplier Sigma Aldrich Supplier No. 110094250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Ethylbutyraldehyde reacts with cyclohexanecarbaldehyde to form homoallylic alcohols.2-Ethylbutyraldehyde has been used in the preparation of aldoximes using aqueous hydroxylamine.

Specifications

CAS 97-96-1
Density 0.814 g/mL (at 25°C (literature))
Boiling Point 117°C (lit.)
Molecular Formula C6H12O
Refractive Index n20/D 1.402 (literature)
Linear Formula (C2H5)2 CHCHO
MDL Number MFCD00006985
Quantity 250 mL
Synonym 2-Ethylbutanal
Molecular Weight (g/mol) 100.16
Percent Purity ≥92%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.