Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylbutyric acid, MP Biomedicals™

Catalog No. ICN15797280 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15797280 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15797280 Supplier MP Biomedicals, Inc Supplier No. 0215797280

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88-09-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
InChI Key OXQGTIUCKGYOAA-UHFFFAOYSA-N
Synonym 2-ethylbutyric acid, diethylacetic acid, butanoic acid, 2-ethyl, acetic acid, diethyl, 3-pentanecarboxylic acid, 2-ethyl butanoic acid, diethyl acetic acid, butyric acid, 2-ethyl, 2-ethyl-butyric acid, alpha-ethylbutyric acid
PubChem CID 6915
IUPAC Name 2-ethylbutanoic acid
SMILES CCC(CC)C(=O)O

Specifications

Color Colorless
Quantity 100 g
Formula Weight 116.16
Physical Form Liquid
Chemical Name or Material 2-Ethylbutyric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.