Learn More
Medchemexpress LLC 2-Ethylfuran-d5 | 2477520-26-4 | 101.16 | 1 MG
SDP
Catalog No. 5000451196
Encompass_Preferred
2-Ethylfuran-d5 is a deuterium labeled compound intended for research use only. It serves as a tracer or an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules for quantitation during drug development.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-51196 Supplier Medchemexpress LLC Supplier No. HYW013015S1MG
2-Ethylfuran-d5 is a deuterium labeled compound intended for research use only. It serves as a tracer or an internal standard for quantitative analysis by methods such as NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements incorporated into drug molecules for quantitation during drug development.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.