Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Ethylhexanal

Catalog No. E291095ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E291095ML 5 mL
E29109250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E291095ML Supplier Sigma Aldrich Supplier No. E291095ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-ethylhexanal is an aliphatic aldehyde used as an intermediate to produce various useful products such as perfumes, disinfectants, paints, warning agents, insecticides, and leak detectors.2-ethylhexanal can be used as:
  • A reductant in the mukaiyama epoxidation of cis-cyclooctene
  • A reactant in the horner-wadsworth-emmons reaction to synthesize cis-α,α-unsaturated amides
  • A starting material to synthesize 2-ethylhexanoic acid using manganese(ii) acetate as a catalyst

Specifications

CAS 123-05-7
Density 0.822 g/mL (at 25°C (literature))
Boiling Point 55°C (13.5 mmHg)
Molecular Formula C8H16O
Refractive Index n20/D 1.415 (literature)
Linear Formula CH3(CH2)3 CH(C2H5)CHO
Quantity 5 mL
MDL Number MFCD00006987
Synonym 2-Ethylcapronaldehyde
Molecular Weight (g/mol) 128.21
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.