Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Ethylhexanoic acid

Catalog No. 538701100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-870-1100ML 100 mL
53-870-11L 1 L
53-870-120L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 53-870-1100ML Supplier Sigma Aldrich Supplier No. 538701100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-ethylhexanoic acid (ehxa, 2-eha) is an industrially important aliphatic carboxylic acid. it is widely employed as a stabilizer and a wood preservative.it has various industrial applications, such as:
  • Coolant in automotives
  • Synthetic lubricant
  • Wetting agent; co-solvent
  • Drying of paints
  • Defoaming agent in pesticides

Specifications

CAS 149-57-5
Density 0.903 g/mL (at 25°C (literature))
Boiling Point 228°C (lit.)
Molecular Formula C8H16O2
Linear Formula CH3(CH2)3 CH(C2H5)CO2H
Refractive Index n20/D 1.425 (literature)
MDL Number MFCD00002675
Quantity 100 mL
Synonym 2-Ethylcaproic acid
Molecular Weight (g/mol) 144.21
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.