Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Ethylhexanoic acid

Catalog No. E291412L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E291412L 2 L
E291411L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E291412L Supplier Sigma Aldrich Supplier No. E291412L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-ethylhexanoic acid can be used:
  • As a reactant in esterification , decarboxylative alkynylation , and preparation of alkyl coumarins via decarboxylative coupling reactions
  • In the organocatalytic medium for the preparation of various 3,4-dihydropyrimidin-2(1h)-ones/thiones by biginelli reaction

Specifications

CAS 149-57-5
Density 0.903 g/mL (at 25°C (literature))
Boiling Point 228°C (lit.)
Molecular Formula C8H16O2
Refractive Index n20/D 1.425 (literature)
Linear Formula CH3(CH2)3 CH(C2H5)CO2H
MDL Number MFCD00002675
Quantity 2 L
Synonym 2-Ethylcaproic acid
Molecular Weight (g/mol) 144.21
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.