Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexanoic acid, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013186 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3186 1 mL
11-101-3187 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3186 Supplier MilliporeSigma Supelco Supplier No. 025291MLF
Add to Cart
Add to Cart

2-Ethylhexanoic acid is one of the di(2-ethylhexyl)phthalate (DEHP) metabolites.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 699°F
CAS 149-57-5
Vapor Pressure 10 mmHg (115°C)
Vapor Density 4.98 (vs. air)
Density 0.903 g/mL (at 25°C)
Flash Point 114°C - Closed Cup
Linear Formula CH3(CH2)3 CH(C2H5)CO2 H
MDL Number MFCD00002675
Boiling Point 228°C
Synonym 2-Ethylcaproic acid
Type Analytical Standard
Quantity 1 mL
Formula Weight 144.21
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.