Learn More
Medchemexpress LLC 2-ethylhexyl 2,3,4,5-tetrabromobenzoate | 183658-27-7 | MFCD26960592 | >=95.0% | 549.92 g·mol⁻¹ | C15H18Br4O2 | 5 MG
SDP
Catalog No. 5000282392
Encompass_Preferred
2-Ethylhexyl 2,3,4,5-tetrabromobenzoate is a brominated benzoate compound used as a flame retardant in polymer and material research. It is intended for research use and is characterized by formula C15H18Br4O2 and molecular weight 549.92 g·mol⁻¹.
Catalog No. 50-002-82392 Supplier Medchemexpress LLC Supplier No. HYW3378865MG
2-Ethylhexyl 2,3,4,5-tetrabromobenzoate is a brominated benzoate compound used as a flame retardant in polymer and material research. It is intended for research use and is characterized by formula C15H18Br4O2 and molecular weight 549.92 g·mol⁻¹.