Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexyl 4-dimethylaminobenzoate, 99%

Catalog No. AAH54136AP
Click to view available options
Quantity:
100 mL
500 mL

CAS: 21245-02-3 | C17H27NO2 | 277.408 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21245-02-3
Molecular Formula C17H27NO2
Molecular Weight (g/mol) 277.408
MDL Number MFCD00017526
InChI Key WYWZRNAHINYAEF-UHFFFAOYSA-N
Synonym 2-ethylhexyl 4-dimethylamino benzoate, padimate o, escalol 507, padimate-o, octyl dimethyl paba, arlatone uvb, benzoic acid, 4-dimethylamino-, 2-ethylhexyl ester, 2-ethylhexyl p-dimethylaminobenzoate, eusolex 6007, sundown
PubChem CID 30541
IUPAC Name 2-ethylhexyl 4-(dimethylamino)benzoate
SMILES CCCCC(CC)COC(=O)C1=CC=C(C=C1)N(C)C

Specifications

Density 0.995
Boiling Point 325°C
Flash Point 113°C (235°F)
Refractive Index 1.542
Quantity 500 mL
Merck Index 14,3232
Formula Weight 277.4
Percent Purity 99%
Chemical Name or Material 2-Ethylhexyl 4-dimethylaminobenzoate
missing translation for 'healthHazard1' H360
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P201-P202-P281-P308+P313-P501c
EINECSNumber 244-289-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.