Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexyl 4-methoxycinnamate, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014565 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4565 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4565 Supplier MilliporeSigma Supelco Supplier No. 55529100MG
Add to Cart
Add to Cart

2-Ethylhexyl 4-methoxycinnamate is a commonly used chemical UV filter, which can generally find applications as a sunscreen ingredient.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 5466-77-3
Flash Point 193°C
Physical Form Neat
Linear Formula C18H26O3
MDL Number MFCD00072582
Synonym 4-Methoxycinnamic acid 2-ethylhexyl ester; OMC; Octyl methoxycinnamate
Type Analytical Standard
Quantity 100 mg
Formula Weight 290.40
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.