Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexyl acetate, 99%, pure

Catalog No. AC410105000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC410105000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC410105000 Supplier Thermo Scientific Chemicals Supplier No. 410105000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 103-09-3 | C10H20O2 | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-09-3
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00027249
InChI Key WOYWLLHHWAMFCB-UHFFFAOYNA-N
Synonym acetic acid, 2-ethylhexyl ester, 2-ethyl-1-hexanol acetate, 2-ethylhexyl ethanoate, 2-ethyl-1-hexyl acetate, 2-ethylhexanyl acetate, 2-ethylhexylacetate, beta-ethylhexyl acetate, acetic acid 2-ethylhexyl ester, fema number 2806, 2-ethylhexylester kyseliny octove
PubChem CID 7635
ChEBI CHEBI:87392
IUPAC Name 2-ethylhexyl acetate
SMILES CCCCC(CC)COC(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -45°C
Color Colorless
Density 0.8700g/mL
Boiling Point 197°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4190 to 1.421
Linear Formula CH3COOCH2CH(C2H5)(CH2)3CH3
Quantity 500 g
Specific Gravity 0.87
Merck Index 15, 6852
Solubility Information Solubility in water: insoluble
Formula Weight 172.27
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl acetate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
EINECSNumber 203-079-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.