Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Ethylhexyl acrylate

Catalog No. 2908153L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 L
3 L
18 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-081-53L 3 L
29-081-518L 18 L
29-081-51L 1 L
29-081-525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 29-081-53L Supplier Sigma Aldrich Supplier No. 2908153L
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-ethylhexyl acrylate (EHA) is one of the constituents of acrylic polymeric latex. It can undergo free-radical solution polymerization. EHA is known to undergo free radical emulsion co- polymerization with methyl methacrylate (MMA) to form pressure-sensitive adhesives. It may also form copolymers with acrylonitrile and acrylamide.

Specifications

CAS 103-11-7
Density 0.885 g/mL (at 25°C)
Boiling Point 215°C to 219°C
Molecular Formula C11H20O2
Refractive Index n20/D 1.436 (literature)
Linear Formula CH2=CHCOOCH2CH(C2H5)(CH2)3 CH3
Quantity 3 L
MDL Number MFCD00009495
Synonym (+/-)-Acrylic acid 2-ethylhexyl ester
Molecular Weight (g/mol) 184.28
Percent Purity 98%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.