Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexyl Butyrate 98.0+%, TCI America™
SDP

Catalog No. B0130500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0130500ML 500 mL
B013025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0130500ML Supplier TCI America Supplier No. B0130500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25415-84-3
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.32
MDL Number MFCD00059428
InChI Key LFEQNZNCKDNGRM-UHFFFAOYNA-N
Synonym Butyric Acid Octyl Ester, Butyric Acid 2-Ethylhexyl Ester, Octyl Butyrate
PubChem CID 117230
IUPAC Name 2-ethylhexyl butanoate
SMILES CCCCC(CC)COC(=O)CCC

Specifications

Color Colorless
Boiling Point 237°C
Quantity 500 mL
Formula Weight 200.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl Butyrate
EINECSNumber (2)-2488
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.