Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexyl cyanoacetate, 98+%, Thermo Scientific™

Catalog No. AC243170250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC243170250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC243170250 Supplier Thermo Scientific Chemicals Supplier No. 243170250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13361-34-7
Molecular Formula C11H19NO2
Molecular Weight (g/mol) 197.28
MDL Number MFCD00019843
InChI Key ZNYBQVBNSXLZNI-UHFFFAOYNA-N
Synonym 2-ethylhexyl cyanoacetate, acetic acid, cyano-, 2-ethylhexyl ester, cyanoacetic acid, 2-ethylhexyl ester, 2-ethylhexylcyanoacetate, cyanoacetic acid octyl ester, acetic acid, 2-cyano-, 2-ethylhexyl ester, cyanoacetic acid 2-ethylhexyl ester, ehca, thcn-nva, acmc-209bt2
PubChem CID 96359
IUPAC Name 2-ethylhexyl 2-cyanoacetate
SMILES CCCCC(CC)COC(=O)CC#N

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless
Density 0.94g/mL
Infrared Spectrum Authentic
Assay Percent Range 98+%
Refractive Index 1.4375 to 1.4395 (20°C, 589nm)
Linear Formula NCCH2COOCH2CH(C2H5)(CH2)3CH3
Quantity 25 mL
Formula Weight 197.28
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl cyanoacetate, 98+%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.