Learn More
Medchemexpress LLC 2-Ethylhexyl diphenyl phosphate | 1241-94-7 | 94.3% | 362.41 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW009066
Encompass_Preferred
2-Ethylhexyl diphenyl phosphate is an organophosphate flame retardant (OPFR) and a PPARG agonist (EC20: 2.04 μM). It also inhibits ERRγ transcriptional activity (IC50: 1.3 μM). This compound upregulates 3β-HSD1, human chorionic gonadotropin (hCG), and progesterone secretion. It can be used in studies of female reproduction and fetal development.
  • Organophosphate flame retardant (OPFR)
  • PPARG agonist (EC20: 2.04 μM)
  • Inhibits ERRγ transcriptional activity (IC50: 1.3 μM)
  • Upregulates 3β-HSD1
  • Upregulates human chorionic gonadotropin (hCG)
  • Upregulates progesterone secretion
  • Suitable for studies of female reproduction and fetal development
Catalog No. 50-004-53466