Learn More
Medchemexpress LLC 2-Ethylhexyl diphenyl phosphate | 1241-94-7 | 94.3% | 362.41 | 50 MG
SDP
Catalog No. 5000453116
Encompass_Preferred
2-Ethylhexyl diphenyl phosphate is an organophosphate flame retardant (OPFR) and acts as a PPARG agonist with an EC20 of 2.04 μM. It also demonstrates inhibitory activity on ERRγ transcriptional activity with an IC50 of 1.3 μM. This compound has been shown to upregulate 3β-HSD1, human chorionic gonadotropin (hCG), and progesterone secretion. It can be utilized in studies pertaining to female reproduction and fetal development.
  • Organophosphate flame retardant (OPFR).
  • PPARG agonist (EC20: 2.04 μM).
  • Inhibits ERRγ transcriptional activity (IC50: 1.3 μM).
  • Upregulates 3β-HSD1, human chorionic gonadotropin (hCG), and progesterone secretion.
  • Applicable for studies in female reproduction and fetal development.
Catalog No. 50-004-53116 Supplier Medchemexpress LLC Supplier No. HYW00906650MG
2-Ethylhexyl diphenyl phosphate is an organophosphate flame retardant (OPFR) and acts as a PPARG agonist with an EC20 of 2.04 μM. It also demonstrates inhibitory activity on ERRγ transcriptional activity with an IC50 of 1.3 μM. This compound has been shown to upregulate 3β-HSD1, human chorionic gonadotropin (hCG), and progesterone secretion. It can be utilized in studies pertaining to female reproduction and fetal development.
  • Organophosphate flame retardant (OPFR).
  • PPARG agonist (EC20: 2.04 μM).
  • Inhibits ERRγ transcriptional activity (IC50: 1.3 μM).
  • Upregulates 3β-HSD1, human chorionic gonadotropin (hCG), and progesterone secretion.
  • Applicable for studies in female reproduction and fetal development.