Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (2-Ethylhexyl)Magnesium Bromide Solution

Catalog No. 630551100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-055-1100ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 63-055-1100ML Supplier Sigma Aldrich Supplier No. 630551100ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(2-Ethylhexyl)magnesium bromide (Grignard reagent) can be used in the synthesis of: 3-ethyl-N,N-dimethyl-1-(pyridin-2-yl)heptan-1-amine, a pyridyl amine ligand, which is used in the polymerization of olefinsDiphenylbis(2-ethylhexyl) germane from diphenylgermanium dichloride by Grignard addition reaction Dichloro (dodecyl) 2-ethylhexylsilane, a key intermediate that is used in the preparation of silafluorene-based copolymers.

Specifications

CAS 112981-69-8
Density 1.015 g/mL at 25°C
Molecular Formula C7H4BrF6Mg
Linear Formula C7H4BrF6Mg
MDL Number MFCD07369723
Quantity 50 mL
Molecular Weight (g/mol) 306.31 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.