Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Ethylhexyl nitrate

Catalog No. 501807218
Encompass
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-180-7218 2.5 L
29-378-41L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-180-7218 Supplier Sigma Aldrich Supplier No. 2937842.5L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Biodegradation of 2-ethylhexyl nitrate (2-ehn) by mycobacterium austroafricanum ifp 2173 has been studied
  • Biodegradation of 2-ehn was studied in biphasic liquid cultures using an inert non-aqueous phase liquid such as 2,2,4,4,6,8,8-heptamethylnonane as solvent.2-ethylhexyl nitrate (2-ehn) is a major fuel additive, has been used to increase the cetane number of diesel
TRUSTED_SUSTAINABILITY

Specifications

CAS 27247-96-7
Density 0.963 g/mL (at 25°C (literature))
Molecular Formula C8H17NO3
Refractive Index n20/D 1.432 (literature)
Linear Formula CH3(CH2)3 CH(C2H5)CH2ONO2
MDL Number MFCD00011582
Quantity 2.5 L
Molecular Weight (g/mol) 175.23
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.