Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Ethylhexyl salicylate

Catalog No. 437379100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-737-9100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-737-9100ML Supplier Sigma Aldrich Supplier No. 437379100ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Ethylhexyl salicylate(EHS) is an organic ultraviolet(UV) absorber that can be used as a photostable ingredient in cosmetic formulations. It shows an absorption spectra in the range of 280-320 nm in the UV region.EHS is a salicylic ester which can be used as an UV filter in sunscreen based creams.

Specifications

CAS 118-60-5
Density 1.014 g/mL (at 25°C)
Boiling Point 189°C to 190°C (21 mmHg)
Molecular Formula C15H22O3
Refractive Index n20/D 1.502 (literature)
Linear Formula (HO)C6H4CO2CH2CH(C2H5)(CH2)3 CH3
Quantity 100 mL
MDL Number MFCD00053300
Synonym Octisalate; Octyl salicylate
Molecular Weight (g/mol) 250.33
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.