Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylhexylamine Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. E012825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E012825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E012825G Supplier TCI America Supplier No. E012825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26392-49-4
Molecular Formula C8H20ClN
Molecular Weight (g/mol) 165.705
MDL Number MFCD00060201
InChI Key JNWRGTDFPPYRAD-UHFFFAOYSA-N
Synonym 1-Amino-2-ethylhexane Hydrochloride, Octylamine Hydrochloride
PubChem CID 117308
IUPAC Name 2-ethylhexan-1-amine;hydrochloride
SMILES CCCCC(CC)CN.Cl

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 165.71
Percent Purity ≥97.0% (N,T)
Physical Form Wax
Chemical Name or Material 2-Ethylhexylamine Hydrochloride
EINECSNumber (2)-0133
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.