Learn More
eMolecules​ 282727-19-9 | 2-Ethylphenylzinc iodide, 0.50 M in THF | Synthonix | MFCD01311456 | 297.440 | C8H9IZn | 0.000 | [I-].CCc1ccccc1[Zn+] | 25ml | 627353629
Catalog No. 502695339 Shop All eMolecules Inc. Products
Encompass_Preferred
2-Ethylphenylzinc iodide, 0.50 M in THF | Synthonix | 282727-19-9 | MFCD01311456 | 297.440 | C8H9IZn | 0.000 | [I-].CCc1ccccc1[Zn+] | 25ml | 627353629
Catalog No. 50-269-5339 Supplier eMolecules​ Supplier No. 627353629
2-Ethylphenylzinc iodide, 0.50 M in THF | Synthonix | 282727-19-9 | MFCD01311456 | 297.440 | C8H9IZn | 0.000 | [I-].CCc1ccccc1[Zn+] | 25ml | 627353629