Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylpyrrole, 98%, Thermo Scientific Chemicals

Catalog No. p-3757904
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC308770010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC308770010 Supplier Thermo Scientific Chemicals Supplier No. 308770010
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1551-06-0
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.15
MDL Number MFCD00060477
InChI Key XRPDDDRNQJNHLQ-UHFFFAOYSA-N
PubChem CID 137075
IUPAC Name 2-ethyl-1H-pyrrole
SMILES CCC1=CC=CN1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 0.9290g/mL
Boiling Point 142.0°C (15.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4985 to 1.5005
Quantity 1 g
Specific Gravity 0.929
Synonym 2-ethylpyrrole, 1h-pyrrole, 2-ethyl, pyrrole,2-ethyl, 1h-pyrrole,2-ethyl, acmc-209dbz, #, 2-ethylpyrrole, analytical standard, 2-ethylpyrrole, technical grade
Formula Weight 95.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Ethylpyrrole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.