Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2-ethylsulfonylphenyl)-[(2S)-4-(6-fluoro-1,3-benzothiazol-2-yl)-2-methylpiperazin-1-yl]methanone | 1971937-18-4 | 99.8% | 447.55 | C21H22FN3O3S2 | 5 MG
SDP

Catalog No. 5000221433
Encompass_Preferred

ARN19702 is a selective, orally active, reversible, and brain-penetrant N-acylethanolamine acid amidase (NAAA) inhibitor. It inhibits human NAAA with an IC50 of 230 nM and has demonstrated analgesic (pain-relief) effects in animal models. Supplied as a white to off-white solid for preclinical research.

  • Selective, orally active reversible NAAA inhibitor.
  • Human NAAA IC50: 230 nM.
  • Brain-penetrant compound suitable for in vivo studies.
  • High purity (99.8%) and white to off-white appearance.
  • Molecular formula C21H22FN3O3S2; molecular weight 447.55.
  • Recommended storage: powder at -20°C for long term or 4°C for short term.

Catalog No. 50-002-21433 Supplier Medchemexpress LLC Supplier No. HY1453395MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ARN19702 is a selective, orally active, reversible, and brain-penetrant N-acylethanolamine acid amidase (NAAA) inhibitor. It inhibits human NAAA with an IC50 of 230 nM and has demonstrated analgesic (pain-relief) effects in animal models. Supplied as a white to off-white solid for preclinical research.

  • Selective, orally active reversible NAAA inhibitor.
  • Human NAAA IC50: 230 nM.
  • Brain-penetrant compound suitable for in vivo studies.
  • High purity (99.8%) and white to off-white appearance.
  • Molecular formula C21H22FN3O3S2; molecular weight 447.55.
  • Recommended storage: powder at -20°C for long term or 4°C for short term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.