Learn More
Medchemexpress LLC (2-ethylsulfonylphenyl)-[(2S)-4-(6-fluoro-1,3-benzothiazol-2-yl)-2-methylpiperazin-1-yl]methanone | 1971937-18-4 | 99.8% | C21H22FN3O3S2 | 10MG
SDP
Catalog No. 5000260278
Encompass_Preferred
ARN19702 is a selective, orally active, reversible, brain-penetrant inhibitor of N-acylethanolamine acid amidase (NAAA) with an IC50 of 230 nM against human NAAA. It has demonstrated analgesic effects in preclinical models and is supplied at high purity for use as a biochemical tool compound in NAAA-related research.
  • Selective N-acylethanolamine acid amidase (NAAA) inhibition for targeted pathway studies.
  • Orally active and brain-penetrant properties enable central nervous system investigations.
  • Reversible mechanism suitable for transient modulation of enzyme activity.
  • Potent activity (IC50 230 nM) for use in cellular and in vivo assays.
  • High purity for reliable biochemical and pharmacological experiments.
Catalog No. 50-002-60278 Supplier Medchemexpress LLC Supplier No. HY14533910MG
ARN19702 is a selective, orally active, reversible, brain-penetrant inhibitor of N-acylethanolamine acid amidase (NAAA) with an IC50 of 230 nM against human NAAA. It has demonstrated analgesic effects in preclinical models and is supplied at high purity for use as a biochemical tool compound in NAAA-related research.
  • Selective N-acylethanolamine acid amidase (NAAA) inhibition for targeted pathway studies.
  • Orally active and brain-penetrant properties enable central nervous system investigations.
  • Reversible mechanism suitable for transient modulation of enzyme activity.
  • Potent activity (IC50 230 nM) for use in cellular and in vivo assays.
  • High purity for reliable biochemical and pharmacological experiments.