Learn More
eMolecules​ 2-Ethylsulfonylphenylboronic acid | 1042443-60-6 | 1G | Purity: 97%
Supplier:  eMolecules​ 117525817
Encompass_Preferred
Combi-Blocks | 2-Ethylsulfonylphenylboronic acid | 1G | 1042443-60-6 | MFCD03095198
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool
Catalog No. 50-158-3326