Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(ethylthio)-1H-benzimidazole | 14610-11-8 | MFCD00223154 | ≥98.0% | 178.25 g/mol | C9H10N2S | 250 MG
SDP

Catalog No. 5000304068
Encompass_Preferred

Bemethyl (2-(ethylthio)-1H-benzimidazole, CAS 14610-11-8) is a synthetic adaptogen supplied as a research-grade powder and used to study antihypoxic and endurance-related biological effects. It is provided for laboratory research and analytical applications and is not intended for human or veterinary use.

  • Synthetic adaptogen and antihypoxant useful for research studies.
  • Chemical name: 2-(ethylthio)-1H-benzimidazole.
  • CAS number: 14610-11-8.
  • Chemical formula C9H10N2S and molecular weight 178.25 g/mol.
  • Available as powder in small research quantities, including 250 mg.
  • Purity ≥98% as reported by suppliers.
  • Recommended storage: powder at -20°C for long term; in solvent at -80°C for extended storage.
  • For research use only; not for human or veterinary use.

Catalog No. 50-003-04068 Supplier Medchemexpress LLC Supplier No. 250MGHYW030562
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Bemethyl (2-(ethylthio)-1H-benzimidazole, CAS 14610-11-8) is a synthetic adaptogen supplied as a research-grade powder and used to study antihypoxic and endurance-related biological effects. It is provided for laboratory research and analytical applications and is not intended for human or veterinary use.

  • Synthetic adaptogen and antihypoxant useful for research studies.
  • Chemical name: 2-(ethylthio)-1H-benzimidazole.
  • CAS number: 14610-11-8.
  • Chemical formula C9H10N2S and molecular weight 178.25 g/mol.
  • Available as powder in small research quantities, including 250 mg.
  • Purity ≥98% as reported by suppliers.
  • Recommended storage: powder at -20°C for long term; in solvent at -80°C for extended storage.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.