Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Ethylthio)ethanol 98.0+%, TCI America™
SDP

Catalog No. E024825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E024825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E024825ML Supplier TCI America Supplier No. E024825ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 110-77-0
Melting Point -100°C
Color Yellow
Boiling Point 182°C
Molecular Formula C4H10OS
Quantity 25 mL
MDL Number MFCD00002909
UN Number 1760
Synonym 2-ethylthio ethanol, ethyl 2-hydroxyethyl sulfide, ethanol, 2-ethylthio, ethyl thioethanol, 2-hydroxyethyl ethyl sulfide, ethyl 2-hydroxyethyl thioether, beta-ethylthioethanol, 2-ethylsulfanyl ethanol, beta-hydroxydiethyl sulfide, 2-ethylthioethanol
InChI Key LNRIEBFNWGMXKP-UHFFFAOYSA-N
SMILES CCSCCO
IUPAC Name 2-(ethylsulfanyl)ethan-1-ol
Molecular Weight (g/mol) 106.18
PubChem CID 8075
Formula Weight 106.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Ethylthio)ethanol
Show More Show Less
EINECSNumber (2)-2452
RTECSNumber KL1225000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.