Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylthiophene 97.0+%, TCI America™
SDP

Catalog No. E038025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E038025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E038025ML Supplier TCI America Supplier No. E038025ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 872-55-9
Molecular Formula C6H8S
Molecular Weight (g/mol) 112.19
MDL Number MFCD00005461
InChI Key JCCCMAAJYSNBPR-UHFFFAOYSA-N
Synonym thiophene, 2-ethyl, 2-ethyl-thiophene, ethylthiophene, thiophene, ethyl, pubchem5536, acmc-209qjc, 2-ethylthiophene, 2-ethylthiophene, analytical standard, met 2-cl-4-metpyrimidine-6-carboxylate
PubChem CID 13388
IUPAC Name 2-ethylthiophene
SMILES CCC1=CC=CS1

Specifications

Color Yellow
Boiling Point 137°C
Quantity 25 mL
UN Number 1993
Formula Weight 112.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.