Learn More
Accela Chembio Inc MF: C7H5N | MW: 103.12 | >97% | Flash Point: 76 celsius | BP: 90¿C/30mmHg | Density: 1.0190.
Catalog No. 50648599
Encompass_Preferred
MF: C7H5N | MW: 103.12 | >97% | Flash Point: 76 celsius | BP: 90¿C/30mmHg | Density: 1.0190.
Catalog No. 50-648-599 Supplier Accela Chembio Inc Supplier No. SY0192725G
MF: C7H5N | MW: 103.12 | >97% | Flash Point: 76 celsius | BP: 90¿C/30mmHg | Density: 1.0190.