Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Ethynylpyridine

Catalog No. 4699201G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-992-01G 1 g
46-992-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-992-01G Supplier Sigma Aldrich Supplier No. 4699201G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-ethynylpyridine is a pyridine derivative.2-ethynylpyridine has been used to synthesize:
  • Poly(2-ethynyl-n-iodopyridinium iodide), via in-situ uncatalyzed polymerization
  • Poly[2-ethynyl-n-(2-furoyl)pyridinium chloride
  • Poly[2-ethynyl-n-(propylsulfonate)pyridinium betaine]
  • 4-(2α-pyridyl)-1,2,3-triphospholide anion, via reaction with anionic heptaphosphide clusters
  • Poly[n-(carboxymethyl)-2-ethynylpyridinium bromide], a new water-soluble ionic conjugated polymer
  • Self-doped ionic conjugated polymer, poly(2-ethynylpyridinimum n-benzoylsulfonate).it may darken in storage with no loss in purity

Specifications

CAS 1945-84-2
Density 1.021 g/mL (at 25°C (literature))
Boiling Point 85°C (12 mmHg)
Molecular Formula C7H5N
Refractive Index n20/D 1.56 (literature)
Linear Formula C7H5N
MDL Number MFCD00041598
Quantity 1 g
Molecular Weight (g/mol) 103.12
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.