Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-F-Bz-dC phosphoramidite | 161442-19-9 | 99.48% | 851.90 | 250 MG
SDP

Catalog No. 5000349373
Encompass_Preferred

2'-F-Bz-dC Phosphoramidite is used in the synthesis of oligoribonucleotides, such as DNA and RNA, and acts as an antiviral agent inhibiting viral replication. It contains a phosphorothioate backbone, which is utilized for synthesizing antisense oligonucleotide analogs to induce apoptosis in cancer cells. These oligonucleotides are short-chain nucleic acids that silence specific genes by complementary base pairing, thereby inhibiting the expression of various proteins.

  • Used in oligoribonucleotide synthesis
  • Acts as an antiviral agent
  • Contains a phosphorothioate backbone
  • Induces apoptosis in cancer cells
  • Silences specific genes
  • Inhibits protein expression
  • Storage at 4°C, protected from light, and under nitrogen
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen

Catalog No. 50-003-49373 Supplier Medchemexpress LLC Supplier No. 250MGHY138577
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-F-Bz-dC Phosphoramidite is used in the synthesis of oligoribonucleotides, such as DNA and RNA, and acts as an antiviral agent inhibiting viral replication. It contains a phosphorothioate backbone, which is utilized for synthesizing antisense oligonucleotide analogs to induce apoptosis in cancer cells. These oligonucleotides are short-chain nucleic acids that silence specific genes by complementary base pairing, thereby inhibiting the expression of various proteins.

  • Used in oligoribonucleotide synthesis
  • Acts as an antiviral agent
  • Contains a phosphorothioate backbone
  • Induces apoptosis in cancer cells
  • Silences specific genes
  • Inhibits protein expression
  • Storage at 4°C, protected from light, and under nitrogen
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.