Learn More
Medchemexpress LLC 2'-F-Bz-dC Phosphoramidite | 161442-19-9 | 99.5% | 851.90 | 100 MG
SDP
Catalog No. 5000349677
Encompass_Preferred
2'-F-Bz-dC Phosphoramidite is a chemical compound used in the synthesis of oligoribonucleotides, such as DNA and RNA. It also serves as an antiviral agent to inhibit virus replication and contains a phosphorothioate backbone for synthesizing antisense oligonucleotide analogs, which can induce apoptosis in cancer cells.
  • Used in the synthesis of oligoribonucleotides (DNA and RNA)
  • Used as an antiviral agent to inhibit virus replication
  • Contains a phosphorothioate backbone for synthesizing antisense oligonucleotide analogs
  • Induces apoptosis in cancer cells
Catalog No. 50-003-49677 Supplier Medchemexpress LLC Supplier No. HY138577100MG
2'-F-Bz-dC Phosphoramidite is a chemical compound used in the synthesis of oligoribonucleotides, such as DNA and RNA. It also serves as an antiviral agent to inhibit virus replication and contains a phosphorothioate backbone for synthesizing antisense oligonucleotide analogs, which can induce apoptosis in cancer cells.
  • Used in the synthesis of oligoribonucleotides (DNA and RNA)
  • Used as an antiviral agent to inhibit virus replication
  • Contains a phosphorothioate backbone for synthesizing antisense oligonucleotide analogs
  • Induces apoptosis in cancer cells