Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-1,4-dimethoxybenzene, 97%, Thermo Scientific Chemicals

Catalog No. AAL1350506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1350506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1350506 Supplier Thermo Scientific Chemicals Supplier No. L1350506
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 82830-49-7 | Molecular Formula: C8H9FO2 | Molecular Weight: 156.156 g/mol

2-Fluoro-1,4-dimethoxybenzene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-1,4-dimethoxybenzene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82830-49-7
Molecular Formula C8H9FO2
Molecular Weight (g/mol) 156.156
MDL Number MFCD00042487
InChI Key WNCYZVMZKSOPMU-UHFFFAOYSA-N
Synonym 1,4-dimethoxy-2-fluorobenzene, 2,5-dimethoxyfluorobenzene, 2,5-dimethoxy-1-fluorobenzene, benzene, 2-fluoro-1,4-dimethoxy, pubchem3050, acmc-209pql, 3-fluoro-1,4-dimethoxybenzene, benzene,2-fluoro-1,4-dimethoxy, 2-fluoranyl-1,4-dimethoxy-benzene, 2-fluorohydroquinone dimethyl ether
PubChem CID 145002
IUPAC Name 2-fluoro-1,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)OC)F

Specifications

Melting Point 23°C to 26°C
Boiling Point 119°C to 121°C (40 mmHg)
Refractive Index 1.5045
Quantity 5 g
Beilstein 6387735
Solubility Information Insoluble in water.
Formula Weight 156.16
Percent Purity 97%
Chemical Name or Material 2-Fluoro-1,4-dimethoxybenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.