Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-1,4-dimethoxybenzene 97.0+%, TCI America™
SDP

Catalog No. F06335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F06335G Supplier TCI America Supplier No. F06335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 82830-49-7
Molecular Formula C8H9FO2
Molecular Weight (g/mol) 156.156
MDL Number MFCD00042487
InChI Key WNCYZVMZKSOPMU-UHFFFAOYSA-N
Synonym 1,4-dimethoxy-2-fluorobenzene, 2,5-dimethoxyfluorobenzene, 2,5-dimethoxy-1-fluorobenzene, benzene, 2-fluoro-1,4-dimethoxy, pubchem3050, acmc-209pql, 3-fluoro-1,4-dimethoxybenzene, benzene,2-fluoro-1,4-dimethoxy, 2-fluoranyl-1,4-dimethoxy-benzene, 2-fluorohydroquinone dimethyl ether
PubChem CID 145002
IUPAC Name 2-fluoro-1,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)OC)F

Specifications

Boiling Point 121°C
Quantity 5 g
Formula Weight 156.16
Percent Purity ≥97.0% (GC)
Chemical Name or Material 2-Fluoro-1,4-dimethoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.