Learn More
Medchemexpress LLC 2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite | 1404463-12-2 | 99.8% | 851.90 g·mol⁻¹ | C46H51FN5O8P | 10 MG
SDP
Catalog No. 5000285857
Encompass_Preferred
2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite is a protected cytidine phosphoramidite monomer intended for use in automated solid-phase oligonucleotide synthesis. Its 2'-fluoro arabino configuration and benzoyl protection facilitate incorporation of modified cytidine residues into synthetic oligonucleotides for research applications in nucleic acid modification, therapeutic oligonucleotides, and epigenetic studies.
  • Suitable for automated solid-phase oligonucleotide synthesis.
  • Enables incorporation of 2'-fluoro arabino cytidine into oligonucleotides.
  • Benzoyl-protected nucleobase improves stability during synthesis.
  • High purity supports consistent coupling efficiency.
  • Supplied in a small research pack for method development and optimization.
  • White to off-white solid form, easy to handle and store.
Catalog No. 50-002-85857 Supplier Medchemexpress LLC Supplier No. HY15441410MG
2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite is a protected cytidine phosphoramidite monomer intended for use in automated solid-phase oligonucleotide synthesis. Its 2'-fluoro arabino configuration and benzoyl protection facilitate incorporation of modified cytidine residues into synthetic oligonucleotides for research applications in nucleic acid modification, therapeutic oligonucleotides, and epigenetic studies.
  • Suitable for automated solid-phase oligonucleotide synthesis.
  • Enables incorporation of 2'-fluoro arabino cytidine into oligonucleotides.
  • Benzoyl-protected nucleobase improves stability during synthesis.
  • High purity supports consistent coupling efficiency.
  • Supplied in a small research pack for method development and optimization.
  • White to off-white solid form, easy to handle and store.