Learn More
Medchemexpress LLC 2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite | 1404463-12-2 | C46H51FN5O8P | 1 ML
SDP
Catalog No. 5000376861
Encompass_Preferred
2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite is a cytidine analog. Cytidine analogs work by inhibiting DNA methyltransferases and show potential anti-metabolic and anti-tumor activities.
  • Cytidine analog
  • Inhibits DNA methyltransferases
  • Shows potential anti-metabolic activities
  • Shows potential anti-tumor activities
Catalog No. 50-003-76861 Supplier Medchemexpress LLC Supplier No. HY15441410MM/1ML
2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite is a cytidine analog. Cytidine analogs work by inhibiting DNA methyltransferases and show potential anti-metabolic and anti-tumor activities.
  • Cytidine analog
  • Inhibits DNA methyltransferases
  • Shows potential anti-metabolic activities
  • Shows potential anti-tumor activities