Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite | 1404463-12-2 | C46H51FN5O8P | 100 MG
SDP

Catalog No. 5000376234
Encompass_Preferred

2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite is a cytidine analog. Cytidine analogs operate by inhibiting DNA methyltransferases, such as Zebularine. These compounds demonstrate potential anti-metabolic and anti-tumor activities. The product is intended for research use only.

  • Purity: 99.84%
  • Appearance: Solid
  • Color: White to off-white
  • Solubility: 125 mg/mL in DMSO (requires ultrasonic treatment; hygroscopic DMSO can affect solubility)
  • Storage: Powder: -20°C for 3 years; In solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-003-76234 Supplier Medchemexpress LLC Supplier No. HY154414100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite is a cytidine analog. Cytidine analogs operate by inhibiting DNA methyltransferases, such as Zebularine. These compounds demonstrate potential anti-metabolic and anti-tumor activities. The product is intended for research use only.

  • Purity: 99.84%
  • Appearance: Solid
  • Color: White to off-white
  • Solubility: 125 mg/mL in DMSO (requires ultrasonic treatment; hygroscopic DMSO can affect solubility)
  • Storage: Powder: -20°C for 3 years; In solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.