Learn More
Medchemexpress LLC 2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite | 1404463-12-2 | C46H51FN5O8P | 25 MG
SDP
Catalog No. 5000375267
Encompass_Preferred
2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite is a cytidine analog. Cytidine analogs work by inhibiting DNA methyltransferases and show potential anti-metabolic and anti-tumor activities. This product is intended for research use only.
  • Cytidine analog
  • Inhibits DNA methyltransferases
  • Shows potential anti-metabolic activity
  • Shows potential anti-tumor activity
  • For research use only
Catalog No. 50-003-75267 Supplier Medchemexpress LLC Supplier No. 25MGHY154414
2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite is a cytidine analog. Cytidine analogs work by inhibiting DNA methyltransferases and show potential anti-metabolic and anti-tumor activities. This product is intended for research use only.
  • Cytidine analog
  • Inhibits DNA methyltransferases
  • Shows potential anti-metabolic activity
  • Shows potential anti-tumor activity
  • For research use only