Learn More
Medchemexpress LLC 2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite | 1404463-12-2 | C46H51FN5O8P | 250 MG
SDP
Catalog No. 5000374436
Encompass_Preferred
2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite is a cytidine analog that works by inhibiting DNA methyltransferases. This compound shows potential anti-metabolic and anti-tumor activities.
  • Cytidine analog
  • Inhibits DNA methyltransferases
  • Possesses potential anti-metabolic activities
  • Possesses potential anti-tumor activities
  • High purity (99.84%)
  • Appears as a white to off-white solid
  • Stable under recommended storage conditions
  • For research use only
Catalog No. 50-003-74436 Supplier Medchemexpress LLC Supplier No. 250MGHY154414
2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite is a cytidine analog that works by inhibiting DNA methyltransferases. This compound shows potential anti-metabolic and anti-tumor activities.
  • Cytidine analog
  • Inhibits DNA methyltransferases
  • Possesses potential anti-metabolic activities
  • Possesses potential anti-tumor activities
  • High purity (99.84%)
  • Appears as a white to off-white solid
  • Stable under recommended storage conditions
  • For research use only