Learn More
Medchemexpress LLC 2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite | 1404463-12-2 | C46H51FN5O8P | 50 MG
SDP
Catalog No. 5000376568
Encompass_Preferred
2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite is a cytidine analog. Cytidine analogs inhibit DNA methyltransferases and possess potential anti-metabolic and anti-tumor activities.
  • Cytidine analog
  • Inhibits DNA methyltransferases
  • Possesses anti-metabolic activity
  • Possesses anti-tumor activity
  • Nucleoside antimetabolite/analog
  • Related to oligonucleotides
  • Related to phosphoramidites
Catalog No. 50-003-76568 Supplier Medchemexpress LLC Supplier No. HY15441450MG
2'-Fluoro-2'-deoxy-ara-C(Bz)-3'-phosphoramidite is a cytidine analog. Cytidine analogs inhibit DNA methyltransferases and possess potential anti-metabolic and anti-tumor activities.
  • Cytidine analog
  • Inhibits DNA methyltransferases
  • Possesses anti-metabolic activity
  • Possesses anti-tumor activity
  • Nucleoside antimetabolite/analog
  • Related to oligonucleotides
  • Related to phosphoramidites