Learn More
Cayman Chemical 2-Fluoro-2-deoxy-arabIno-Ur 1g
SDP
Catalog No. 502942190
Encompass_Preferred
An antiviral nucleoside derivative; inhibits HSV-1 replication in infected Vero cells at 1 UG/ml; not cytotoxic to L5178Y or P815 cells (IC50 = >10 UG/ml for both)
Catalog No. 50-294-2190 Supplier Cayman Chemical Supplier No. 403031 G
An antiviral nucleoside derivative; inhibits HSV-1 replication in infected Vero cells at 1 UG/ml; not cytotoxic to L5178Y or P815 cells (IC50 = >10 UG/ml for both)