Learn More
Cayman Chemical 2'-Fluoro-2'-deoxy-arabino-Uridine, Purity: ≥98%, CAS: 69123-94-0, 5 g
SDP
Catalog No. 502942365
Encompass_Preferred
An antiviral nucleoside derivative; inhibits HSV-1 replication in infected Vero cells at 1 µg/ml; not cytotoxic to L5178Y or P815 cells (IC50 = >10 µg/ml for both)
Catalog No. 50-294-2365 Supplier Cayman Chemical Supplier No. 403035 G
An antiviral nucleoside derivative; inhibits HSV-1 replication in infected Vero cells at 1 µg/ml; not cytotoxic to L5178Y or P815 cells (IC50 = >10 µg/ml for both)