Learn More
AdipoGen 2-Fluoro-2-deoxy-D-glucose
SDP
Supplier:  AdipoGen CDXF0076M100
Encompass_Preferred
Chemical. CAS 29702-43-0. Formula C6H11FO5. MW 182.15. 2-Fluoro-2-deoxy-D-glucose is a glucose analog that inhibits cellular glycosylation. It is the natural isotope-form of 18F-FDG that can be taken up by cells but does not undergo metabolic glycolysis. FDG uptake is closely related to the expression of the glucose transporter GLUT in malignant tumours. FDG is also a substrate of hexokinase isozymes and an inhibitor of cellular glycosylation. FDG is taken up by high-glucose-using cells such as brain, brown adipocytes, kidney and cancer cells, where phosphorylation prevents the glucose from being released again from the cell, once it has been absorbed. The 2-hydroxyl group –OH in normal glucose is needed for further glycolysis metabolism of glucose by splitting it, but FDG is missing this 2-hydroxyl and cannot be further metabolized in cells. FDG is used in positron emission tomography PET as contrast agent and is the most widely used PET marker. FDG-PET can be used fo
Catalog No. 50-256-0900